7-Bromo-1-methylbenzodiazole 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Bromo-1-methyl-1H-benzo[d]imidazole
OR910284-10G
OR910284-1G
OR910284-5G
APOSOR910284-10GEA
475
EUR
APOSOR910284-10G
APOSOR910284-1G
APOSOR910284-5G
7-Bromo-1-methylbenzodiazole 98%
7-Bromo-1-methylbenzodiazole
Formel:
C₈H₇BrN₂ Molecular Weight: 211.059 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135915 CAS-Nummer: 1233542-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...