7-Bromo-1-methylbenzodiazole 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-1-methyl-1H-benzo[d]imidazole

OR910284-10G OR910284-1G OR910284-5G
APOSOR910284-10GEA 475 EUR
APOSOR910284-10G APOSOR910284-1G APOSOR910284-5G
7-Bromo-1-methylbenzodiazole 98%
7-Bromo-1-methylbenzodiazole
Formel: C₈H₇BrN₂
Molecular Weight: 211.059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135915
CAS-Nummer: 1233542-00-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR