2-Benzyl-5-methyl-4H-pyrazol-3-one 97%

Lieferant: Apollo Scientific

OR910288-1G
APOSOR910288-1GEA 270 EUR
APOSOR910288-1G
2-Benzyl-5-methyl-4H-pyrazol-3-one 97%
2-Benzyl-5-methyl-4H-pyrazol-3-one
Formel: C₁₁H₁₂N₂O
Molecular Weight: 188.226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00461211
CAS-Nummer: 946-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR