1-(Ethanesulphonyl)pyrrole 98%

Lieferant: Apollo Scientific

OR910308-1G
APOSOR910308-1GEA 319 EUR
APOSOR910308-1G
1-(Ethanesulphonyl)pyrrole 98%
1-(Ethanesulphonyl)pyrrole
Formel: C₆H₉NO₂S
Molecular Weight: 159,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05144864
CAS-Nummer: 885440-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR