3-(3,5-Dimethylpyrazol-1-yl)benzonitrile 98%
Lieferant: Apollo Scientific
OR910315-1G
APOSOR910315-1GEA
270
EUR
APOSOR910315-1G
3-(3,5-Dimethylpyrazol-1-yl)benzonitrile 98%
3-(3,5-Dimethylpyrazol-1-yl)benzonitrile
Formel:
C₁₂H₁₁N₃ Molecular Weight: 197,236 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12134526 CAS-Nummer: 1155066-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...