3-(5-Hydroxy-3-methylpyrazol-1-yl)propanenitrile 98%
Lieferant: Apollo Scientific
OR910325-1G
APOSOR910325-1GEA
319
EUR
APOSOR910325-1G
3-(5-Hydroxy-3-methylpyrazol-1-yl)propanenitrile 98%
3-(5-Hydroxy-3-methylpyrazol-1-yl)propanenitrile
Formel:
C₇H₉N₃O Molecular Weight: 151,166 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00634617 CAS-Nummer: 41534-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...