6-Chloro-3-methyl-1,2-benzoxazole 98%

Lieferant: Apollo Scientific

OR910367-1G
APOSOR910367-1GEA 400 EUR
APOSOR910367-1G
6-Chloro-3-methyl-1,2-benzoxazole 98%
6-Chloro-3-methyl-1,2-benzoxazole
Formel: C₈H₆ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD07778389
CAS-Nummer: 66033-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR