7-chloro-1,3-benzoxazol-2-amine 95%

Lieferant: Apollo Scientific

Synonyme: 7-Chlorobenzo[d]oxazol-2-amine

OR910370-1G
APOSOR910370-1GEA 369 EUR
APOSOR910370-1G
7-chloro-1,3-benzoxazol-2-amine 95%
7-chloro-1,3-benzoxazol-2-amine
Formel: C₇H₅ClN₂O
Molecular Weight: 168,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01664219
CAS-Nummer: 64037-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR