5-Nitro-1H-indazol 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR9104-100G
OR9104-500G
OR9104-25G
APOSOR9104-100GEA
69.6
EUR
APOSOR9104-100G
APOSOR9104-500G
APOSOR9104-25G
5-Nitro-1H-indazol 95%
5-Nitro-1H-indazole
Formel:
C₇H₅N₃O₂ Schmelzpunkt: 204…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005693 CAS-Nummer: 5401-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...