Ethyl 2-(4-methylimidazol-1-yl)acetate 95%
Lieferant: Apollo Scientific
OR910439-1G
APOSOR910439-1GEA
719
EUR
APOSOR910439-1G
Ethyl 2-(4-methylimidazol-1-yl)acetate 95%
Ethyl 2-(4-methylimidazol-1-yl)acetate
Formel:
C₈H₁₂N₂O₂ Molecular Weight: 168,193 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21336474 CAS-Nummer: 6338-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...