5-Methoxy-1-methyl-1,3-benzodiazole 96%

Lieferant: Apollo Scientific

Synonyme: 5-Methoxy-1-methyl-1H-benzo[d]imidazole

OR910453-1G
APOSOR910453-1GEA 66.7 EUR
APOSOR910453-1G
5-Methoxy-1-methyl-1,3-benzodiazole 96%
5-Methoxy-1-methyl-1,3-benzodiazole
Formel: C₉H₁₀N₂O
Molecular Weight: 162,188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00962665
CAS-Nummer: 10394-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR