4,6-Dimethoxyindol 98%

Lieferant: Apollo Scientific

OR910472-1G
APOSOR910472-1GEA 502 EUR
APOSOR910472-1G
4,6-Dimethoxyindol 98%
4,6-Dimethoxyindole
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128213
CAS-Nummer: 23659-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR