Oxindol-6-carbonsäure 97%

Lieferant: Apollo Scientific

Synonyme: 2-Oxoindoline-6-carboxylic acid

OR910478-5G
APOSOR910478-5GEA 95.7 EUR
APOSOR910478-5G
Oxindol-6-carbonsäure 97%
Oxindol-6-carbonsäure
Formel: C₉H₇NO₃
Molecular Weight: 177.1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01415801
CAS-Nummer: 334952-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR