Methyl indoline-7-carboxylate 98%

Lieferant: Apollo Scientific

OR910525-1G OR910525-250MG
APOSOR910525-1GEA 230 EUR
APOSOR910525-1G APOSOR910525-250MG
Methyl indoline-7-carboxylate 98%
Methyl indoline-7-carboxylate
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954772
CAS-Nummer: 112106-91-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR