((R)-Piperidin-2-yl)methanol 95%

Lieferant: Apollo Scientific

Synonyme: (R)-Piperidin-2-ylmethanol

OR910571-1G
APOSOR910571-1GEA 88.6 EUR
APOSOR910571-1G
((R)-Piperidin-2-yl)methanol 95%
((R)-Piperidin-2-yl)methanol
Formel: C₆H₁₃NO
Molecular Weight: 115,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11036292
CAS-Nummer: 3197-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR