1,6-Dimethylindazol-3-amine 98%
Lieferant: Apollo Scientific
OR910573-1G
APOSOR910573-1GEA
319
EUR
APOSOR910573-1G
1,6-Dimethylindazol-3-amine 98%
1,6-Dimethylindazol-3-amine
Formel:
C₉H₁₁N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22549327 CAS-Nummer: 1276113-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...