tert-Butyl 3-bromoindazole-1-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-bromo-1H-indazole-1-carboxylate

OR910593-1G
APOSOR910593-1GEA 226 EUR
APOSOR910593-1G
tert-Butyl 3-bromoindazole-1-carboxylate 97%
tert-Butyl 3-bromoindazole-1-carboxylate
Formel: C₁₂H₁₃BrN₂O₂
Molecular Weight: 297,148 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22380231
CAS-Nummer: 1257296-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR