3-methoxy-5-nitrobenzonitrile 97%

Lieferant: Apollo Scientific

OR910730-5G
APOSOR910730-5GEA 128 EUR
APOSOR910730-5G
3-methoxy-5-nitrobenzonitrile 97%
3-methoxy-5-nitrobenzonitrile
Formel: C₈H₆N₂O₃
Molecular Weight: 178.145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463876
CAS-Nummer: 33224-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR