3-(methylamino)benzonitrile 95%
Lieferant: Apollo Scientific
Synonyme:
3-(Methylamino)benzonitrile
OR910959-1G
OR910959-250MG
APOSOR910959-1GEA
89.9
EUR
APOSOR910959-1G
APOSOR910959-250MG
3-(methylamino)benzonitrile 95%
3-(methylamino)benzonitrile
Formel:
C₈H₈N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11153024 CAS-Nummer: 64910-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...