Salicylaldoxim 98%

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxybenzaldehyde oxime

OR911122-500G
APOSOR911122-500GEA 232 EUR
APOSOR911122-500G
Salicylaldoxim 98%
Salicylaldoxim
Formel: C₇H₇NO₂
Molecular Weight: 137.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002120
CAS-Nummer: 94-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR