2-Hydroxy-3-methoxybenzaldehyde oxime 98%

Lieferant: Apollo Scientific

Synonyme: 2-[(1E)-(hydroxyimino)methyl]-6-methoxyphenol

OR911127-1G
APOSOR911127-1GEA 77 EUR
APOSOR911127-1G
2-Hydroxy-3-methoxybenzaldehyde oxime 98%
2-Hydroxy-3-methoxybenzaldehyde oxime
Formel: C₈H₉NO₃
Molecular Weight: 167.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01043210
CAS-Nummer: 2169-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR