3-(4-Ethoxyphenyl)propionsäure 95%

Lieferant: Apollo Scientific

Synonyme: 3-(4-Ethoxyphenyl)propanoic acid

OR911154-25G
APOSOR911154-25GEA 243 EUR
APOSOR911154-25G
3-(4-Ethoxyphenyl)propionsäure 95%
3-(4-Ethoxyphenyl)propionsäure
Formel: C₁₁H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00016557
CAS-Nummer: 4919-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR