2-Isopropoxy-6-methylpyridine 96%

Lieferant: Apollo Scientific

OR911168-1G
APOSOR911168-1GEA 364 EUR
APOSOR911168-1G
2-Isopropoxy-6-methylpyridine 96%
2-Isopropoxy-6-methylpyridine
Formel: C₉H₁₃NO
Molecular Weight: 151,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23114609
CAS-Nummer: 1395039-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR