(3-Propoxyphenyl)acetic acid 98%

Lieferant: Apollo Scientific

OR911179-1G OR911179-250MG
APOSOR911179-1GEA 328 EUR
APOSOR911179-1G APOSOR911179-250MG
(3-Propoxyphenyl)acetic acid 98%
(3-Propoxyphenyl)acetic acid
Formel: C₁₁H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07754771
CAS-Nummer: 900704-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR