3-cyano-4-methoxybenzoic acid 95%
Lieferant: Apollo Scientific
OR911190-1G
APOSOR911190-1GEA
122
EUR
APOSOR911190-1G
3-cyano-4-methoxybenzoic acid 95%
3-cyano-4-methoxybenzoic acid
Formel:
C₉H₇NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09271826 CAS-Nummer: 117738-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...