3-cyano-4-ethoxybenzoic acid 95%

Lieferant: Apollo Scientific

Synonyme: 3-Cyano-4-ethoxybenzoic acid

OR911191-1G
APOSOR911191-1GEA 398 EUR
APOSOR911191-1G
3-cyano-4-ethoxybenzoic acid 95%
3-cyano-4-ethoxybenzoic acid
Formel: C₁₀H₉NO₃
Molecular Weight: 191,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09271828
CAS-Nummer: 258273-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR