2-Methoxy-3-methylaniline 97%

Lieferant: Apollo Scientific
Achtung

OR911198-1G OR911198-5G
APOSOR911198-1GEA 75.4 EUR
APOSOR911198-1G APOSOR911198-5G
2-Methoxy-3-methylaniline 97%
2-Methoxy-3-methylaniline
Formel: C₈H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD12547890
CAS-Nummer: 18102-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR