7-Methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR9112-1G
APOSOR9112-1GEA
306
EUR
APOSOR9112-1G
7-Methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid
7-Methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylic acid
Formel:
C₁₅H₁₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269534 CAS-Nummer: 220465-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...