4-(N-methylacetamido)benzoic acid 97%

Lieferant: Apollo Scientific

OR911241-1G
APOSOR911241-1GEA 400 EUR
APOSOR911241-1G
4-(N-methylacetamido)benzoic acid 97%
4-(N-methylacetamido)benzoic acid
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20542852
CAS-Nummer: 26961-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR