3-Chloro-1H-quinolin-2-one 95%

Lieferant: Apollo Scientific

OR911269-1G
APOSOR911269-1GEA 538 EUR
APOSOR911269-1G
3-Chloro-1H-quinolin-2-one 95%
3-Chloro-1H-quinolin-2-one
Formel: C₉H₆ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD08705671
CAS-Nummer: 939-17-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR