L-Methioninmethylester Hydrochlorid 96%
Lieferant: Apollo Scientific
Synonyme:
H-Met-OMe.HCl, H-Met-OMe·HCl
OR911324-100G
APOSOR911324-100GEA
84.1
EUR
APOSOR911324-100G
L-Methioninmethylester Hydrochlorid 96%
L-Methioninmethylester Hydrochlorid
Formel:
C₆H₁₃NO₂S·HCl Molecular Weight: 199,7 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012491 CAS-Nummer: 2491-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...