1-Benzyl 3-methyl piperidine-1,3-dicarboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 1-Cbz-piperidine-3-carboxylate

OR911373-100G
APOSOR911373-100GEA 74.1 EUR
APOSOR911373-100G
1-Benzyl 3-methyl piperidine-1,3-dicarboxylate 98%
1-Benzyl 3-methyl piperidine-1,3-dicarboxylate
Formel: C₁₅H₁₉NO₄
Molecular Weight: 277.316 g/mol
Schmelzpunkt: 273…278 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06795953
CAS-Nummer: 174543-74-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR