trans-4-Isopropylcyclohexancarbonsäure 98%

Lieferant: Apollo Scientific

OR911376-100G OR911376-25G OR911376-500G
APOSOR911376-100GEA 0 EUR
APOSOR911376-100G APOSOR911376-25G APOSOR911376-500G
trans-4-Isopropylcyclohexancarbonsäure 98%
trans-4-Isopropylcyclohexancarbonsäure
Formel: C₁₀H₁₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04004145
CAS-Nummer: 7077-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR