(1H,3H)-Chinazolin-2,4-dion 98%

Lieferant: Apollo Scientific

Synonyme: Quinazoline-2,4(1H,3H)-dione, Benzoyleneurea

OR911378-100G
APOSOR911378-100GEA 174 EUR
APOSOR911378-100G
(1H,3H)-Chinazolin-2,4-dion 98%
(1H,3H)-Chinazolin-2,4-dion
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006699
CAS-Nummer: 86-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR