Ethyl-m-anisat 98%

Lieferant: Apollo Scientific

Synonyme: m-Anissäureethylester, Ethyl-3-methoxybenzoat, 3-Methoxybenzoesäureethylester, 3-Anissäureethylester

OR911401-100G
APOSOR911401-100GEA 97.3 EUR
APOSOR911401-100G
Ethyl-m-anisat 98%
Ethyl-m-anisat
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00015149
CAS-Nummer: 10259-22-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR