3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-on 98%

Lieferant: Apollo Scientific

Synonyme: 1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal)

OR911428-100G
APOSOR911428-100GEA 127 EUR
APOSOR911428-100G
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-on 98%
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-on
Formel: C₁₁H₁₈O₃
Molecular Weight: 198.26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006652
CAS-Nummer: 69225-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR