D-Tyrosine methyl ester hydrochloride 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-D-Tyr-OMe.HCl, H-D-Tyr-OMe·HCl

OR911434-100G OR911434-500G OR911434-25G
APOSOR911434-100GEA 111 EUR
APOSOR911434-100G APOSOR911434-500G APOSOR911434-25G
D-Tyrosine methyl ester hydrochloride 98+%
D-Tyrosine methyl ester hydrochloride
Formel: C₁₀H₁₄ClNO₃
Molecular Weight: 231.676 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066124
CAS-Nummer: 3728-20-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR