3-(3-Nitrophenyl)-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
OR9115-1G
OR9115-250MG
APOSOR9115-1GEA
46.4
EUR
APOSOR9115-1G
APOSOR9115-250MG
3-(3-Nitrophenyl)-1H-pyrazole
3-(3-Nitrophenyl)-1H-pyrazole
Formel:
C₉H₇N₃O₂ Schmelzpunkt: 126…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00106146 CAS-Nummer: 59843-77-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...