3-Hydroxybenzylalkohol 97%

Lieferant: Apollo Scientific

OR911508-100G
APOSOR911508-100GEA 168 EUR
APOSOR911508-100G
3-Hydroxybenzylalkohol 97%
3-Hydroxybenzylalkohol
Formel: C₇H₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00004643
CAS-Nummer: 620-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR