2-Hydroxyquinoline-4-carboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxycinchoninic Acid

OR911510-500G
APOSOR911510-500GEA 183 EUR
APOSOR911510-500G
2-Hydroxyquinoline-4-carboxylic acid 98%
2-Hydroxyquinoline-4-carboxylic acid
Formel: C₁₀H₇NO₃
Molecular Weight: 189.17 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023942
CAS-Nummer: 15733-89-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR