3-(1-Cyanethyl)benzoesäure 98%
Lieferant: Apollo Scientific
OR911534-100G
APOSOR911534-100GEA
58
EUR
APOSOR911534-100G
3-(1-Cyanethyl)benzoesäure 98%
3-(1-Cyanethyl)benzoesäure
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002520 CAS-Nummer: 5537-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...