3-(Chlormethyl)benzonitril 98%

Lieferant: Apollo Scientific

Synonyme: 3-Cyanobenzyl chloride

OR911554-25G
APOSOR911554-25GEA 72.5 EUR
APOSOR911554-25G
3-(Chlormethyl)benzonitril 98%
3-(Chlormethyl)benzonitril
Formel: C₈H₆ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD02684191
CAS-Nummer: 64407-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR