4-Butylcyclohexanecarboxylic acid (cis and trans mixture) 98%

Lieferant: Apollo Scientific

OR911645-1G OR911645-25G OR911645-5G
APOSOR911645-1GEA 102 EUR
APOSOR911645-1G APOSOR911645-25G APOSOR911645-5G
4-Butylcyclohexanecarboxylic acid (cis and trans mixture) 98%
4-Butylcyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₁₁H₂₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00460757
CAS-Nummer: 71101-89-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR