2-Isopropyl-3-methoxypyrazin 98%

Lieferant: Apollo Scientific

OR911656-25G
APOSOR911656-25GEA 157 EUR
APOSOR911656-25G
2-Isopropyl-3-methoxypyrazin 98%
2-Isopropyl-3-methoxypyrazin
Formel: C₈H₁₂N₂O
Molecular Weight: 152,1900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006135
CAS-Nummer: 25773-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR