3,3-Pentamethylene glutarimide 98%

Lieferant: Apollo Scientific

Synonyme: Gabapentin Impurity 3

OR911683-100G OR911683-25G
APOSOR911683-100GEA 144 EUR
APOSOR911683-100G APOSOR911683-25G
3,3-Pentamethylene glutarimide 98%
3,3-Pentamethylene glutarimide
Formel: C₁₀H₁₅NO₂
Molecular Weight: 181.235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023872
CAS-Nummer: 1130-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR