[3-(4-Nitrophenyl)-1H-pyrazol-1-yl]acetic acid

Lieferant: Apollo Scientific
Achtung

OR9117-500MG
APOSOR9117-500MGEA 0 EUR
APOSOR9117-500MG
[3-(4-Nitrophenyl)-1H-pyrazol-1-yl]acetic acid
[3-(4-Nitrophenyl)-1H-pyrazol-1-yl]acetic acid
Formel: C₁₁H₉N₃O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00665862
CAS-Nummer: 959582-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR