N-Ethyl-4-hydroxypiperidine 97%
Lieferant: Apollo Scientific
OR911703-5G
APOSOR911703-5GEA
69.6
EUR
APOSOR911703-5G
N-Ethyl-4-hydroxypiperidine 97%
N-Ethyl-4-hydroxypiperidine
Formel:
C₇H₁₅NO Molecular Weight: 129.2001 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006509 CAS-Nummer: 3518-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...