Methyl tetrahydrophthalic anhydride 95%

Lieferant: Apollo Scientific

OR911727-5G
APOSOR911727-5GEA 0 EUR
APOSOR911727-5G
Methyl tetrahydrophthalic anhydride 95%
Methyl tetrahydrophthalic anhydride
Formel: C₉H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00870768
CAS-Nummer: 19438-64-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR