(R)-(+)-2-Hydroxy-3-phenylpropionsäure 98%

Lieferant: Apollo Scientific

Synonyme: D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-​(+)​-​Phenyllactic acid

OR911731-100G
APOSOR911731-100GEA 178 EUR
APOSOR911731-100G
(R)-(+)-2-Hydroxy-3-phenylpropionsäure 98%
(R)-(+)-2-Hydroxy-3-phenylpropionsäure
Formel: C₉H₁₀O₃
Molecular Weight: 166.1739 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078062
CAS-Nummer: 7326-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR