Boc-D-ABU-OH 97%

Lieferant: Apollo Scientific

Synonyme: (R)-2-(Boc-amino)butyric acid

OR911736-25G
APOSOR911736-25GEA 68.3 EUR
APOSOR911736-25G
Boc-D-ABU-OH 97%
Boc-D-ABU-OH
Formel: C₉H₁₇NO₄
Molecular Weight: 203.238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00270335
CAS-Nummer: 45121-22-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR